Please login to your online account to display your discounted pricing

SKF 77434 hydrobromide, Tocris

Selective D1-like partial agonist

$133.98 - $567.83

Chemical Identifiers

Chemical Name or Material SKF 77434 hydrobromide
Assay Percent Range >99%
Molecular Formula C19H21NO2·HBr
Formula Weight 376.29
Synonym 2,3,4,5-Tetrahydro-1-phenyl-3-(2-propenyl)-1H-3-benzazepine-7,8-diol hydrobromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

166250

 
r & d systems
166250
50mg Each for $567.83

166210

 
r & d systems
166210
10mg Each for $133.98
Description & Specifications

Specifications

Formula Weight 376.29
Quantity 50mg
Assay Percent Range >99%
Synonym 2,3,4,5-Tetrahydro-1-phenyl-3-(2-propenyl)-1H-3-benzazepine-7,8-diol hydrobromide
Chemical Name or Material SKF 77434 hydrobromide
Molecular Formula C19H21NO2·HBr
CAS 300561-58-4
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C