Please login to your online account to display your discounted pricing

(+)-SK&F 10047 hydrochloride, Tocris

σ1 selective agonist

$252.39 - $252.39

Chemical Identifiers

Chemical Name or Material (+)-SK&F 10047 hydrochloride
Assay Percent Range >98%
Molecular Formula C17H23NO·HCl
Formula Weight 293.84
Synonym [2S-(2α,6α,11R™]-1,2,3,4,5,6-hexahydro-6,11-dimethyl-3-(2-propenyl)-2,6-methano-3-benzazocin-8-ol hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

107910

 
r & d systems
RDS107910
10mg Each for $252.39
Description & Specifications

Specifications

Formula Weight 293.84
Quantity 10mg
Molecular Formula C17H23NO·HCl
Synonym [2S-(2α,6α,11R™]-1,2,3,4,5,6-hexahydro-6,11-dimethyl-3-(2-propenyl)-2,6-methano-3-benzazocin-8-ol hydrochloride
Chemical Name or Material (+)-SK&F 10047 hydrochloride
CAS 133005-41-1
Assay Percent Range >98%
Recommended Storage Desiccate at +4°C