Please login to your online account to display your discounted pricing

SJ 172550, Tocris™

MDMX inhibitor; Disrupts MDMX-p53 interaction

$214.12 - $895.76

Chemical Identifiers

Chemical Name or Material SJ 172550
Assay Percent Range >99%
Molecular Formula C22H21ClN2O5
Formula Weight 428.87
Synonym 2-[2-Chloro-4-[(1, 5-dihydro-3-methyl-5-oxo-1-phenyl-4H-pyrazol-4-ylidene)methyl]-6-ethoxyphenoxy]acetic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

392910

 
r & d systems
RDS392910
10mg Each for $214.12

392950

 
r & d systems
RDS392950
50mg Each for $895.76
Description & Specifications

Specifications

Chemical Name or Material SJ 172550
Quantity 10mg
CAS 431979-47-4
Recommended Storage Store at +4°C
Assay Percent Range >99%
Molecular Formula C22H21ClN2O5
Formula Weight 428.87
Synonym 2-[2-Chloro-4-[(1, 5-dihydro-3-methyl-5-oxo-1-phenyl-4H-pyrazol-4-ylidene)methyl]-6-ethoxyphenoxy]acetic acid methyl ester
Safety and Handling

Store at +4°C