Please login to your online account to display your discounted pricing

Sivelestat sodium salt, Tocris™

Selective leukocyte elastase inhibitor

$139.08 - $574.46

Chemical Identifiers

Chemical Name or Material Sivelestat sodium salt
Assay Percent Range >98%
Molecular Formula C20H21N2NaO7S
Formula Weight 456.44
Synonym N-{2-[({4-[(2, 2-Dimethylpropanoyl)oxy]phenyl}sulfonyl)amino]benzoyl}glycine sodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

353550

 
r & d systems
RDS353550
50mg Each for $574.46

353510

 
r & d systems
RDS353510
10mg Each for $139.08
Description & Specifications

Specifications

Formula Weight 456.44
Quantity 50mg
Assay Percent Range >98%
Recommended Storage Desiccate at Room Temperature
Molecular Formula C20H21N2NaO7S
Synonym N-{2-[({4-[(2, 2-Dimethylpropanoyl)oxy]phenyl}sulfonyl)amino]benzoyl}glycine sodium salt
Chemical Name or Material Sivelestat sodium salt
CAS 150374-95-1
Safety and Handling

Desiccate at Room Temperature