Please login to your online account to display your discounted pricing

Sirtinol, Tocris™

Selective sirtuin family deacetylase inhibitor

$218.74 - $915.10

Chemical Identifiers

Chemical Name or Material Sirtinol
Assay Percent Range >98%
Molecular Formula C26H22N2O2
Formula Weight 394.47
Synonym 2-[[(2-Hydroxy-1-naphthalenyl)methylene]amino]-N-(1-phenylethyl)benzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

352110

 
r & d systems
RDS352110
10mg Each for $218.74

352150

 
r & d systems
RDS352150
50mg Each for $915.10
Description & Specifications

Specifications

Formula Weight 394.47
Quantity 10mg
Assay Percent Range >98%
CAS 410536-97-9
Synonym 2-[[(2-Hydroxy-1-naphthalenyl)methylene]amino]-N-(1-phenylethyl)benzamide
Chemical Name or Material Sirtinol
Molecular Formula C26H22N2O2
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C