missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sipatrigine, Tocris™

NaV blocker; neuroprotective

$257.85 - $1,073.10

Chemical Identifiers

Assay Percent Range >99%
Molecular Formula C15H16Cl3N5
Formula Weight 372.68
Synonym 2-(4-Methyl-1-piperazinyl)-5-(2, 3, 5-trichlorophenyl)-4-pyrimidinamine
Chemical Name or Material Sipatrigine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
404450 R & D Systems
404450
50mg Each for $1,073.10
404410 R & D Systems
404410
10mg Each for $257.85
Specifications Description & Specifications

Chemical Identifiers

>99%
372.68
Sipatrigine
C15H16Cl3N5
2-(4-Methyl-1-piperazinyl)-5-(2, 3, 5-trichlorophenyl)-4-pyrimidinamine

Specifications

130800-90-7
C15H16Cl3N5
2-(4-Methyl-1-piperazinyl)-5-(2, 3, 5-trichlorophenyl)-4-pyrimidinamine
Sipatrigine
>99%
372.68
50mg

Safety and Handling

Recommended Storage : Store at Room Temperature