Please login to your online account to display your discounted pricing

Sino Biological™ XPNPEP2 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human XPNPEP2 (O43895) (Met 1-Ala 650), without the pro peptide, fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS11903H08H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11903H08H50

  • / Each
 

Description & Specifications

Specifications

Format Lyophilized
Validated Application Assay Standard
Protein Subtype Proteases
Protein Tag His Tag
Molecular Weight 72kDa
Gene Alias RP4-753P9.2, APP2
Endotoxin Concentration <1 EU/μg
Expression System Mammalian
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Family Other Proteins
Gene Symbol XPNPEP2
Common Name X-prolyl aminopeptidase (aminopeptidase P) 2, membrane-bound
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Purity ≥90% by SDS-PAGE
Quantity 50μg

X-prolyl aminopeptidase (aminopeptidase P) 2, membrane-bound (XPNPEP2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to cleave the fluorogenic peptide substrate, H-Lys(2-Aminobenzoyl)Pro-Pro-pNitroanilide(K(Abz)PP-pNA); the specific activity is >300pmole/min./μg
  • Formulation: Lyophilized in 25mM Tris, 150mM NaCl, pH 7.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.