missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

VCAM1 Recombinant Human Protein, N-term His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human VCAM1 (NP_001069.1) (Phe 25 - Pro 697) fused to a polyhistidine tag at the N-terminus

$275.00 - $275.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias VCAM1, CD106, DKFZp779G2333, INCAM-100, mgC99561
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10113H07H50 View Documents Life Technologies
LS10113H07H50
50μg Each for $275.00
 
Description

Description

Vascular cell adhesion molecule 1 (VCAM1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured by the ability of the immobilized protein to support the adhesion of U937 human histiocytic lymphoma cells; when cells are added to VCAM1 coated plates (10μg/mL, 100μL/well ) approximately 85-95% will adhere specifically
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Lyophilized
VCAM1, CD106, DKFZp779G2333, INCAM-100, mgC99561

For Research Use Only. Not for use in diagnostic procedures.