Please login to your online account to display your discounted pricing

Sino Biological™ VCAM1 Recombinant Human Protein, N-term His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human VCAM1 (NP_001069.1) (Phe 25 - Pro 697) fused to a polyhistidine tag at the N-terminus

Manufacturer: life technologies  LS10113H07H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10113H07H50

  • / Each
 

Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Common Name Vascular cell adhesion molecule 1
Expression System Mammalian
Gene Symbol VCAM1
Protein Family CD Proteins
Validated Application Assay Standard
Gene Alias VCAM1, CD106, DKFZp779G2333, INCAM-100, mgC99561
Protein Tag His Tag
Structural Form Truncated/Recombinant/Cell Surface Protein
Protein Subtype Other Proteins
Endotoxin Concentration <1 EU/μg
Molecular Weight 76kDa
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Quantity 50μg

Vascular cell adhesion molecule 1 (VCAM1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

  • Activity: Measured by the ability of the immobilized protein to support the adhesion of U937 human histiocytic lymphoma cells; when cells are added to VCAM1 coated plates (10μg/mL, 100μL/well ) approximately 85-95% will adhere specifically
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.