missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ USP7 Recombinant Human Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding the amino acid sequence (Lys 208-Glu 560) of human USP7 (NP_003461.2) fused to two additional aa (Gly & Pro) at the N-terminus

$273.00 - $273.00

Specifications

Purity ≥95% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias HAUSP, TEF1
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11681HNCBL Life Technologies
LS11681HNCB250
5 x 50μg Each for $273.00
Specifications

Description

Ubiquitin Carboxyl-Terminal Hydrolase 7 (USP7), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥95% by SDS-PAGE
<1 EU/μg
HAUSP, TEF1
PAGE
Lyophilized
7874