Please login to your online account to display your discounted pricing

Sino Biological™ USP7 Recombinant Human Protein, N-His.GST Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding full length of human USP7 (NP_003461.2) (Met 1-Asn 1102) fused to the N-terminal polyhistidine-tagged GST tag at the N-terminus

$289.00 - $289.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias HAUSP, TEF1
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

11681H20BL2

DFS Item
 
life technologies
LS11681H20BL25
5 x 5μg Each for $289.00
Description & Specifications

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias HAUSP, TEF1

Ubiquitin Carboxyl-Terminal Hydrolase 7 (USP7), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human P53 at 2μg/mL (100μL/well) can bind human USP7 with a linear range of 6.4-160ng/mL
  • Formulation: Lyophilized in 50mM Tris 100mM NaCl, pH 8, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.