missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

USP7 Recombinant Human Protein, N-His.GST Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding full length of human USP7 (NP_003461.2) (Met 1-Asn 1102) fused to the N-terminal polyhistidine-tagged GST tag at the N-terminus

Manufacturer:  Invitrogen LS11681H20BL25

 View more versions of this product

Catalog No. 11681H20BL2



Description

Description

Ubiquitin Carboxyl-Terminal Hydrolase 7 (USP7), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human P53 at 2μg/mL (100μL/well) can bind human USP7 with a linear range of 6.4-160ng/mL
  • Formulation: Lyophilized in 50mM Tris 100mM NaCl, pH 8, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Insect
Proteases
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
156kDa
5 x 5μg
<1 EU/μg
Ubiquitin carboxyl-terminal hydrolase 7
GST Tag, His Tag
Other Proteins
Assay Standard
Lyophilized
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
HAUSP, TEF1
USP7
SDS
Documents

For Research Use Only. Not for use in diagnostic procedures.