missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ UNG Recombinant Human Protein, N-GST Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human UNG isoform 1 (P13051-2) (Phe 85-Leu 304) fused to the GST tag at the N-terminus

$158.00 - $397.00

Specifications

Purity ≥85% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias DGU, DKFZp781L1143, HIGM4, UDG, UNG1, UNG15, UNG2
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
12939H09E25 Life Technologies
LS12939H09E25
5 x 5μg Each for $397.00
12939H09E5 Life Technologies
LS12939H09E5
5μg Each for $158.00
Specifications

Description

Uracil-DNA Glycosylase (UNG), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 40mM Tris, 150mM NaCl, 2mM GSH, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥85% by SDS-PAGE
<1 EU/μg
DGU, DKFZp781L1143, HIGM4, UDG, UNG1, UNG15, UNG2
PAGE
Lyophilized
7374