Please login to your online account to display your discounted pricing

Sino Biological™ UNG Recombinant Human Protein, N-GST Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human UNG isoform 1 (P13051-2) (Phe 85-Leu 304) fused to the GST tag at the N-terminus

Manufacturer: life technologies  LS12939H09E5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 12939H09E5

  • / Each
 

Description & Specifications

Specifications

Purity ≥85% by SDS-PAGE
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Termination N-Terminal
Gene Alias DGU, DKFZp781L1143, HIGM4, UDG, UNG1, UNG15, UNG2
Validated Application Assay Standard
Protein Family Mitochondrial Proteins
Expression System E. coli
Format Lyophilized
Molecular Weight 52kDa
Purification Method PAGE
Quantity 5μg
Structural Form Full Length/Recombinant
Endotoxin Concentration <1 EU/μg
Common Name Uracil-DNA glycosylase
Gene Symbol UNG
Protein Tag GST Tag
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Gene ID (Entrez) 7374

Uracil-DNA Glycosylase (UNG), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 40mM Tris, 150mM NaCl, 2mM GSH, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.