Please login to your online account to display your discounted pricing

Sino Biological™ UNG Recombinant Human Protein, N-GST Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human UNG isoform 1 (P13051-2) (Phe 85-Leu 304) fused to the GST tag at the N-terminus

Manufacturer: life technologies  LS12939H09E25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 12939H09E25

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 52kDa
Purification Method PAGE
Purity ≥85% by SDS-PAGE
Quantity 5 x 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Termination N-Terminal
Structural Form Full Length/Recombinant
Endotoxin Concentration <1 EU/μg
Gene Alias DGU, DKFZp781L1143, HIGM4, UDG, UNG1, UNG15, UNG2
Common Name Uracil-DNA glycosylase
Gene Symbol UNG
Protein Tag GST Tag
Validated Application Assay Standard
Protein Family Mitochondrial Proteins
Expression System E. coli
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 7374

Uracil-DNA Glycosylase (UNG), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 40mM Tris, 150mM NaCl, 2mM GSH, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.