Please login to your online account to display your discounted pricing

Sino Biological™ UCHL3 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human UCHL3 (NP_005993.1) (Glu 2-Ala 230) fused to a polyhistide tag at the N-terminus

$153.00 - $153.00

Specifications

Purity ≥90% by SDS-PAGE
Format Frozen
Gene Alias RP11-173B14.3, UCH-L3, UCHL3
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

11634H07E5

DFS Item
 
life technologies
LS11634H07E5
5μg Each for $153.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Format Frozen
Gene Alias RP11-173B14.3, UCH-L3, UCHL3

Ubiquitin Carboxyl-Terminal Hydrolase Isozyme L3 (UCHL3), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to hydrolyze Ubiquitin AMC; the specific activity is >14,000pmole/min./μg
  • Formulation: 20mM Tris, 500mM NaCl, 0.2μg/μL BSA, 20%glycerol, 1mM DTT pH 8.0
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.