Please login to your online account to display your discounted pricing

Sino Biological™ UCHL3 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human UCHL3 (NP_005993.1) (Glu 2-Ala 230) fused to a polyhistide tag at the N-terminus

Manufacturer: life technologies  LS11634H07E5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11634H07E5

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 27kDa
Quantity 5μg
Format Frozen
Gene Symbol UCHL3
Expression System E. coli
Validated Application Assay Standard
Purity ≥90% by SDS-PAGE
Storage Requirements Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Subtype Proteases
Gene Alias RP11-173B14.3, UCH-L3, UCHL3
Common Name Ubiquitin carboxyl-terminal hydrolase isozyme L3
Protein Tag His Tag
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Termination N-Terminal

Ubiquitin Carboxyl-Terminal Hydrolase Isozyme L3 (UCHL3), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to hydrolyze Ubiquitin AMC; the specific activity is >14,000pmole/min./μg
  • Formulation: 20mM Tris, 500mM NaCl, 0.2μg/μL BSA, 20%glycerol, 1mM DTT pH 8.0
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.