Please login to your online account to display your discounted pricing

Sino Biological™ UCHL1 (UCH-L1) Recombinant Mouse Protein, N-His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mouse UCHL1 (Q9R0P9) (Gln 2-Ala 223) fused to a polyhistide tag at the N-terminus

$122.00 - $122.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Format Frozen/Lyophilized
Gene Alias AW822034, C88048, PGP9.5, R75593, gad
Gene ID (Entrez) 22223
View More Specs
Includes:  Five vials of protein in suspension buffer Store at -80 °C.

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50690M07E25

DFS Item
 
life technologies
LS50690M07E25
5 x 5μg Each for $122.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Format Frozen/Lyophilized
Gene Alias AW822034, C88048, PGP9.5, R75593, gad
Gene ID (Entrez) 22223

Ubiquitin Carboxy-terminal Hydrolase L1 (UCHL1), recombinant mouse protein is supplied frozen. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by the hydrolysis of UbiquitinAMC; the specific activity is >14,000pmole/min./μg
  • Formulation: 50mM Tris, 150mM NaCl, 20% Glycerol, pH 7.7, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.