Please login to your online account to display your discounted pricing

Sino Biological™ UCHL1 Recombinant Human Protein, N-His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the native human UCHL1 (NP_004172.2) (Gln 2-Ala 223) fused to a polyhistide tag at the N-terminus

$152.00 - $152.00

Specifications

Purity ≥90% by SDS-PAGE
Format Frozen
Gene Alias UCHL1, PARK5, PGP 9.5, PGP9.5, PGP95, Uch-L1
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

11663H07E5

DFS Item
 
life technologies
LS11663H07E5
5μg Each for $152.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Format Frozen
Gene Alias UCHL1, PARK5, PGP 9.5, PGP9.5, PGP95, Uch-L1

Ubiquitin Carboxyl-Terminal Hydrolase Isozyme L1 (UCHL1), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to hydrolyze Ubiquitin AMC; the specific activity is >140pmole/min./μg
  • Formulation: 20mM Tris, 500mM NaCl, 0.2μg/μL BSA, 20% glycerol, 1mM DTT, pH 8.0
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.