Please login to your online account to display your discounted pricing

Sino Biological™ UCHL1 Recombinant Human Protein, N-His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the native human UCHL1 (NP_004172.2) (Gln 2-Ala 223) fused to a polyhistide tag at the N-terminus

Manufacturer: life technologies  LS11663H07E5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11663H07E5

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Validated Application Assay Standard
Molecular Weight 25.6kDa
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Quantity 5μg
Format Frozen
Expression System E. coli
Gene Symbol UCHL1
Protein Subtype Proteases
Common Name Ubiquitin carboxyl-terminal hydrolase isozyme L1
Purity ≥90% by SDS-PAGE
Gene Alias UCHL1, PARK5, PGP 9.5, PGP9.5, PGP95, Uch-L1
Protein Tag His Tag

Ubiquitin Carboxyl-Terminal Hydrolase Isozyme L1 (UCHL1), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to hydrolyze Ubiquitin AMC; the specific activity is >140pmole/min./μg
  • Formulation: 20mM Tris, 500mM NaCl, 0.2μg/μL BSA, 20% glycerol, 1mM DTT, pH 8.0
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.