Please login to your online account to display your discounted pricing

Sino Biological™ TXNRD1 Recombinant Human Protein, N-His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human TXNRD1 isoform 1 (Q16881-1) (Tyr 161-Cys 647) fused to a polyhistidinetag at the N-terminus

Manufacturer: life technologies  LS13093H07E50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 13093H07E50

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 7296
Molecular Weight 55kDa
Purification Method PAGE
Purity ≥85% by SDS-PAGE
Quantity 50μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Termination N-Terminal
Structural Form Truncated/Recombinant
Endotoxin Concentration <1 EU/μg
Gene Alias GRIM-12, mgC9145, TR, TR1, TRXR1, TXNR
Common Name Thioredoxin reductase 1
Gene Symbol TXNRD1
Validated Application Assay Standard
Protein Family Other Proteins
Expression System E. coli
Protein Tag His Tag

Thioredoxin Reductase 1 (TXNRD1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.