Please login to your online account to display your discounted pricing

Sino Biological™ TREML1 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human TREML1 isoform 1 (Q86YW5-1) extracellular domain (Met 1-Pro 162) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS11934H08H5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11934H08H5

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Molecular Weight 17.3kDa
Gene ID (Entrez) 340205
Purification Method PAGE
Gene Alias UNQ1825/PRO3438, GLTL1825, mgC119173, PRO3438, TLT-1, TLT1, dJ238O23.3
Protein Subtype Other Proteins
Common Name Triggering receptor expressed on myeloid cells-like 1
Quantity 5μg
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Purity ≥85% by SDS-PAGE
Format Lyophilized
Endotoxin Concentration <1 EU/μg
Structural Form Truncated/Recombinant
Gene Symbol TREML1
Protein Family Other Proteins
Protein Tag His Tag
Termination C-Terminal
Validated Application Assay Standard
Expression System Mammalian

Triggering Receptor Expressed on Myeloid Cells Like 1 (TREML1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.