missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TREM2 Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mouse TREM2 (NP_112544.1) extracellular domain (Met 1-Pro 168) fused to a C-terminal polyhistidine tag

$102.00 - $102.00

Specifications

Purity ≥95% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias Trem2a, Trem2b, Trem2c
View More Specs

Includes: One vial of protein in suspension buffer

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50149M08H5 View Documents Life Technologies
LS50149M08H5
5μg Each for $102.00
 
Description

Description

Triggering Receptor Expressed on Myeloid Cells 2 (TREM2), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥95% by SDS-PAGE
<1 EU/μg
Trem2a, Trem2b, Trem2c
PAGE
Lyophilized
83433

For Research Use Only. Not for use in diagnostic procedures.