missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TREM1 Recombinant Human Protein, hIgG1-Fc.His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Arg 200) of human TREM1 (NP_061113.1) precursor fused to the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus

$102.00 - $221.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias TREM-1, TREM1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10511H03H25 View Documents Life Technologies
LS10511H03H25
5 x 5μg Each for $221.00
 
10511H03H5 View Documents Life Technologies
LS10511H03H5
5μg Each for $102.00
 
Description

Description

Triggering Receptor Expressed on Myeloid Cells 1 (TREM1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
TREM-1, TREM1
PAGE
Lyophilized
54210

For Research Use Only. Not for use in diagnostic procedures.