Please login to your online account to display your discounted pricing

Sino Biological™ TREM1 Recombinant Human Protein, hIgG1-Fc.His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Arg 200) of human TREM1 (NP_061113.1) precursor fused to the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Manufacturer: life technologies  LS10511H03H250

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10511H03HL

  • / Each
 

Description & Specifications

Specifications

Quantity 5 x 50μg
Purity ≥90% by SDS-PAGE
Gene ID (Entrez) 54210
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Protein Tag IgG1 Fc Tag
Molecular Weight 48.3kDa
Gene Symbol TREM1
Expression System Mammalian
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration <1 EU/μg
Purification Method PAGE
Common Name Triggering receptor expressed on myeloid cells 1
Gene Alias TREM-1, TREM1
Structural Form Truncated/Recombinant
Protein Family Other Proteins
Termination C-Terminal
Validated Application Assay Standard
Format Lyophilized

Triggering Receptor Expressed on Myeloid Cells 1 (TREM1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.