Please login to your online account to display your discounted pricing

Sino Biological™ TP53 (P53) Recombinant Cynomolgus Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding the cynomolgus (Macaca fascicularis) P53 (E3U906) (Met 1-Asp 393)

$158.00 - $2,055.00

Specifications

Purity ≥85% by SDS-PAGE
Format Lyophilized
Gene Alias p53, TP53
View More Specs
Includes:  Five vials of lyophilized protein

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

90001CNAE25

DFS Item
 
life technologies
LS90001CNAE25
5 x 5μg Each for $402.00

90001CNAE5

DFS Item
 
life technologies
LS90001CNAE5
5μg Each for $158.00

90001CNAE50

DFS Item
 
life technologies
LS90001CNAE50
50μg Each for $642.00

90001CNAEL

DFS Item
 
life technologies
LS90001CNAE250
5 x 50μg Each for $2,055.00
Description & Specifications

Specifications

Purity ≥85% by SDS-PAGE
Format Lyophilized
Gene Alias p53, TP53

Tumor protein p53 (P53), recombinant cynomolgus protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: 1. Measured by its ability to bind biotinylated human S100B in a functional ELISA; 2. Measured by its ability to bind biotinylated human RRM2B in a functional ELISA
  • Formulation: Lyophilized in 100mM Tris, 3% Glycerol, 4mM DTT, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.