Please login to your online account to display your discounted pricing

Sino Biological™ TNFSRF9 Recombinant Human Protein, hIgG1-Fc.His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the N-terminal fragment (Met 1-Gln 186) of the extracellular domain of human TNFSRF9 (NP_001552.2) fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Manufacturer: life technologies  LS10041H03H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10041H03H50

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 45.2kDa
Purity ≥90% by SDS-PAGE
Quantity 50μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Subtype TNF Receptors
Endotoxin Concentration <1 EU/μg
Gene Alias 4-1BB, CD137, CDw137, ILA, mgC2172, TNFSRF9
Common Name Tumor necrosis factor receptor superfamily, member 9
Gene Symbol TNFSRF9
Protein Tag His Tag/IgG1 Fc Tag
Validated Application Assay Standard
Protein Family Cytokines & Receptors/CD Proteins
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized

Tumor Necrosis Factor Receptor Super Family 9 (TNFSRF9), recombinant human/mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant mouse 4-1BB Ligand at 20μg/mL (100μL/well) can bind human 4-1BB with a linear range of 15.6-500ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.