Please login to your online account to display your discounted pricing

Sino Biological™ TNFSRF9 (CD137) Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gln 186) of human TNFSRF9 (NP_001552.2) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10041H08H25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10041H08H25

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 18.8kDa
Purity ≥85% by SDS-PAGE
Quantity 5 x 5μg
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Structural Form Truncated/Recombinant
Protein Subtype TNF (Tumor Necrosis Factor)
Endotoxin Concentration <1 EU/μg
Gene Alias 4-1BB, CD137, CDw137, ILA, mgC2172, TNFSRF9
Common Name Tumor necrosis factor receptor superfamily, member 9
Gene Symbol TNFSRF9
Protein Tag His Tag
Validated Application Assay Standard
Protein Family Cytokines & Receptors/CD Proteins
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 3604

Tumor necrosis factor receptor superfamily, member 9 (TNFSRF9/CD137), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.