Please login to your online account to display your discounted pricing

Sino Biological™ TNFSF9 Recombinant Mouse Protein, N-His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Arg 104-Glu 309) of the mouse TNFSF9 precursor (NP_033430.1) fused to a polyhistidine tag at the N-terminus

$386.00 - $386.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias TNFSF9, 4-1BB-L, 4-1BBL, AI848817, Cd137l, Ly63l
View More Specs
Includes:  Five vials of lyophilized protein

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50067M07H25

DFS Item
 
life technologies
LS50067M07H25
5 x 5μg Each for $386.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias TNFSF9, 4-1BB-L, 4-1BBL, AI848817, Cd137l, Ly63l

Tumor necrosis factor (ligand) superfamily, member 9 (TNFSF9), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant mouse TNFSF9 at 20μg/mL (100μL/well) can bind human TNFSRF9 with a linear range of 15.6-500ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.