Please login to your online account to display your discounted pricing

Sino Biological™ TNFSF13B Recombinant Human Protein, hIgG1-Fc Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the soluble form of human TNFSF13B (Q9Y275-1) (Ala 134-Leu 285) fused to the Fc region of human IgG1 at the N-terminus

Manufacturer: life technologies  LS10056H01H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10056H01H50

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 43.7kDa
Purity ≥90% by SDS-PAGE
Quantity 50μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Subtype TNF (Tumor Necrosis Factor)
Endotoxin Concentration <1 EU/μg
Gene Alias TNFSF13B, BAFF, BLYS, CD257, DTL, TALL-1, TALL1, THANK, TNFSF20, ZTNF4
Common Name Tumor necrosis factor (ligand) superfamily, member 13b
Gene Symbol TNFSF13B
Protein Tag IgG1 Fc Tag
Validated Application Assay Standard
Protein Family Cytokines & Receptors/CD Proteins
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized

Tumor Necrosis Factor Super Family 13 subunit beta (TNFSF13B), recombinant human/mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human TNFSF13B at 1μg/mL (100μL/well) can bind human TNFRSF17, the EC50 of human TNFRSF17 is 0.07μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.