missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TNFSF10 Recombinant Human Protein, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human TNFSF10 (NP_003801.1) (Val 114-Gly 281) fused to an initial Met

$210.00 - $210.00

Specifications

Purity ≥95% by SDS-PAGE
Format Lyophilized
Gene Alias APO2L, Apo-2L, CD253, TL2, TRAIL
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10409HNAE25 View Documents Life Technologies
LS10409HNAE25
5 x 5μg Each for $210.00
 
Description

Description

Tumor Necrosis (Ligand) Receptor Superfamily Member 10 (TNFSF10), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: 1. Measured in a cytotoxicity assay using L929 mouse fibrosarcoma cells in the presence of the metabolic inhibitor actinomycin D, the ED50 for this effect is typically 4-8ng/mL; 2. Measured in a cytotoxicity assay using NCI-H460 cells in the presence of the metabolic inhibitor actinomycin D, the ED50 for this effect is typically 0.2-0.8ng/mL
  • Formulation: Lyophilized in 40mM Tris 0.3M NaCl pH 7.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥95% by SDS-PAGE
Lyophilized
APO2L, Apo-2L, CD253, TL2, TRAIL

For Research Use Only. Not for use in diagnostic procedures.