Please login to your online account to display your discounted pricing

Sino Biological™ TNFSF10 Recombinant Human Protein DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human TNFSF10 (NP_003801.1) (Val 114-Gly 281) fused to an initial Met

Manufacturer: life technologies  LS10409HNAE25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10409HNAE25

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 19.6kDa
Purity ≥95% by SDS-PAGE
Quantity 5 x 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Subtype TNF (Tumor Necrosis Factor)
Gene Alias APO2L, Apo-2L, CD253, TL2, TRAIL
Common Name Tumor necrosis (ligand) receptor superfamily, member 10
Gene Symbol TNFSF10
Protein Tag Untagged
Validated Application Assay Standard
Protein Family Cytokines & Receptors/CD Proteins
Expression System E. coli
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized

Tumor Necrosis (Ligand) Receptor Superfamily Member 10 (TNFSF10), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

  • Activity: 1. Measured in a cytotoxicity assay using L929 mouse fibrosarcoma cells in the presence of the metabolic inhibitor actinomycin D, the ED50 for this effect is typically 4-8ng/mL; 2. Measured in a cytotoxicity assay using NCI-H460 cells in the presence of the metabolic inhibitor actinomycin D, the ED50 for this effect is typically 0.2-0.8ng/mL
  • Formulation: Lyophilized in 40mM Tris 0.3M NaCl pH 7.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.