Please login to your online account to display your discounted pricing

Sino Biological™ TNFRSF8 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human TNFRSF8 (NP_001234.2) extracellular domain (Met 1-Lys 379) fused to the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus

$199.50 - $199.50

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias TNFRSF8, CD30, D1S166E, Ki-1
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10777H08H25

DFS Item
 
life technologies
LS10777H08H25
5 x 5μg Each for $199.50
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias TNFRSF8, CD30, D1S166E, Ki-1

Tumor Necrosis Factor Receptor Superfamily Member 8 (TNFRSF8), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human TNFRSF8 at 2μg/mL (100μL/well) can bind human CD30L Fc Chimera with a linear range of 6.4-800ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.