missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TNFRSF4 (CD134) Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ala 216) of human TNFRSF4 (NP_003318.1) fused to a polyhistidine tag at the C-terminus

$401.00 - $401.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias TNFRSF4, RP5-902P8.3, ACT35, CD134, OX40, TXGP1L
Gene ID (Entrez) 7293
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10481H08H50 View Documents Life Technologies
LS10481H08H50
50μg Each for $401.00
 
Description

Description

Tumor necrosis factor receptor superfamily, member 4 (TNFRSF4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human TNFRSF4-His at 2μg/mL (100μL/well) can bind human TNFSF4-mouse Fc, the ED50 of human TNFSF4-mouse Fc is 29ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥85% by SDS-PAGE
Lyophilized
7293
<1 EU/μg
TNFRSF4, RP5-902P8.3, ACT35, CD134, OX40, TXGP1L

For Research Use Only. Not for use in diagnostic procedures.