Please login to your online account to display your discounted pricing

Sino Biological™ TNFRSF4 (CD134) Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ala 216) of human TNFRSF4 (NP_003318.1) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10481H08H5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10481H08H5

  • / Each
 

Description & Specifications

Specifications

Endotoxin Concentration <1 EU/μg
Molecular Weight 21.7kDa
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Gene ID (Entrez) 7293
Purity ≥85% by SDS-PAGE
Gene Alias TNFRSF4, RP5-902P8.3, ACT35, CD134, OX40, TXGP1L
Expression System Mammalian
Protein Family Cytokines & Receptors/CD Proteins
Protein Subtype TNF (Tumor Necrosis Factor)
Quantity 5μg
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Gene Symbol TNFRSF4
Structural Form Truncated/Recombinant
Protein Tag His Tag
Common Name Tumor necrosis factor receptor superfamily, member 4
Format Lyophilized
Validated Application Assay Standard

Tumor necrosis factor receptor superfamily, member 4 (TNFRSF4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human TNFRSF4-His at 2μg/mL (100μL/well) can bind human TNFSF4-mouse Fc, the ED50 of human TNFSF4-mouse Fc is 29ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.