missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ TNFRSF21 Recombinant Human Protein, hIgG1-Fc Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Leu 350)of human DR6 (NP_055267.1) precursor fused to Fc region of human IgG1 at the C-terminus

$274.00 - $274.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias BM-018, DR6, mgC31965, TNFRSF21
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10175H02H50 Life Technologies
LS10175H02H50
50μg Each for $274.00
Specifications

Description

Tumor Necrosis Factor Receptor Superfamily, Member 21 (TNFRSF21), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: 1. Measured by its binding ability in a functional ELISA; 2. Immobilized recombinant human DR6-Fc (Mfr. No. 10175-H02H) at 10μg/mL (100μL/well) can bind biotinylated human APP-Fc (Mfr. No. 10703-H02H) with a linear range of 0.03-0.25μg/mL
  • Formulation: Lyophilized in 50mM Tris, 100mM Glycine, 10mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
BM-018, DR6, mgC31965, TNFRSF21
PAGE
Lyophilized
27242