missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ TNFRSF1A (CD120a) Recombinant Mouse Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ala 212) of mouse TNFRSF1A (NP_ 035739.2) fused to a polyhistidine tag at the C-terminus

$245.00 - $245.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias Tnfrsf1, CD120a, FPF, TNF-R, TNF-R-I, TNF-R1, TNF-R55, TNF-alphaR1, TNFAR, TNFR60, TNFRI
Gene ID (Entrez) 21937
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50496M08H25 Life Technologies
LS50496M08H25
5 x 5μg Each for $245.00
Specifications

Description

Tumor necrosis factor receptor superfamily member 1A (TNFRSF1A/CD120a), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.
  • Activity: 1. Measured by its binding ability in a functional ELISA: a. Immobilized human TNFA at 10μg/mL (100μL/well) can bind biotinylated mouse TNFRSF1A-his, the ED50 of biotinylated mouse TNFRSF1A-his is 0.28μg/mL; b. Immobilized mouse TNFA at 10μg/mL (100μL/well) can bind biotinylated mouse TNFRSF1A-his, the ED50 of biotinylated mouse TNFRSF1A-his is 0.13μg/mL; 2. Measured by its ability to inhibit the TNF mediated cytotoxicity in the L929 mouse fibrosarcoma cells in the presence of the metabolic inhibitor actinomycin D, the ED50 for this effect is typically 0.5-1.5μg/mL in the presence of 0.1ng/mL recombinant mouse TNF
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
21937
<1 EU/μg
Tnfrsf1, CD120a, FPF, TNF-R, TNF-R-I, TNF-R1, TNF-R55, TNF-alphaR1, TNFAR, TNFR60, TNFRI