Please login to your online account to display your discounted pricing

Sino Biological™ TNFRSF1A (CD120a) Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Thr 211) of human TNFRSF1A (NP_001056.1) fused to a polyhistidine tag at the C-terminus.

Manufacturer: life technologies  LS10872H08H25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10872H08H25

  • / Each
 

Description & Specifications

Specifications

Expression System Mammalian
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Gene ID (Entrez) 7132
Gene Alias CD120a, FPF, mgC19588, TBP1, TNF-R, TNF-R-I, TNF-R55, TNFAR, TNFR60, p55, p55-R, p60
Protein Family Cytokines & Receptors/CD Proteins
Molecular Weight 22.7kDa
Format Lyophilized
Structural Form Truncated/Recombinant
Quantity 5 x 5μg
Validated Application Assay Standard
Gene Symbol TNFRSF1A
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Protein Tag His Tag
Protein Subtype TNF (Tumor Necrosis Factor)
Common Name Tumor necrosis factor receptor superfamily, member 1A
Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg

Tumor necrosis factor receptor superfamily member 1A (TNFRSF1A/CD120a), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.

  • Activity: 1. Measured by its binding ability in a functional ELISA; a. Immobilized human TNFa at 10μg/mL (100μL/well) can bind biotinylated human TNFRSF1A-his, the ED50 of biotinylated human TNFRSF1A-his is 0.04μg/mL; b. Immobilized mouse TNFa at 10μg/mL (100μL/well) can bind biotinylated human TNFRSF1A-his, the ED50 of biotinylated human TNFRSF1A-his is 0.28μg/mL; 2. Measured by its ability to inhibit the TNF mediated cytotoxicity in the L929 mouse fibrosarcoma cells in the presence of the metabolic inhibitor actinomycin D, the ED50 for this effect is typically 0.04-0.2μg/mL in the presence of 0.25ng/mL recombinant human TNF
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.