missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TNFRSF17 Recombinant Human Protein, hIgG1-Fc.His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human TNFRSF17 (NP_001183.2) (Met 1-Ala 54) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus and a signal peptide at the N-terminus

$225.00 - $366.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias TNFRSF17, BCM, BCMA, CD269
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10620H03H25 View Documents Life Technologies
LS10620H03H25
5 x 5μg Each for $225.00
 
10620H03H50 View Documents Life Technologies
LS10620H03H50
50μg Each for $366.00
 
Description

Description

Tumor necrosis factor receptor superfamily, member 17 (TNFRSF17), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human TNFSF13B/BAFF at 1μg/mL (100μL/well) can bind human TNFRSF17, the EC50 of human TNFRSF17 is 0.07μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥85% by SDS-PAGE
<1 EU/μg
Lyophilized
TNFRSF17, BCM, BCMA, CD269

For Research Use Only. Not for use in diagnostic procedures.