Please login to your online account to display your discounted pricing

Sino Biological™ TNFRSF10D (TRAILR4) Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human TNFRSF10D (NP_003831.2) (Met 1-His 211) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10413H08H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10413H08H50

  • / Each
 

Description & Specifications

Specifications

Protein Subtype TNF Receptors
Validated Application Assay Standard
Gene Alias TNFRSF10D, CD264, DCR2, TRAILR4, TRUNDD
Format Lyophilized
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Molecular Weight 18.4kDa
Gene Symbol TNFRSF10D
Endotoxin Concentration <1 EU/μg
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Quantity 50μg
Protein Tag His Tag
Expression System Mammalian
Protein Family Cytokines & Receptors/CD Proteins
Purity ≥85% by SDS-PAGE
Common Name Tumor necrosis factor receptor superfamily, member 10d, decoy with truncated death domain

Tumor necrosis factor receptor superfamily, member 10D, decoy with truncated death domain (TNFRSF10D), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human TNFRSF10D at 10μg/mL (100μL/well) can bind biotinylated TNFSF10 with a linear range of 0.625-40ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.