Please login to your online account to display your discounted pricing

Sino Biological™ TNFRSF10D (TRAILR4) Recombinant Human Protein, hIgG1-Fc.His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-His 211) of human TNFRSF10D (NP_003831.2) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

$97.50 - $846.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias TNFRSF10D, CD264, DCR2, TRAILR4, TRUNDD
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10413H03H25

DFS Item
 
life technologies
LS10413H03H25
5 x 5μg Each for $216.00

10413H03H50

DFS Item
 
life technologies
LS10413H03H50
50μg Each for $280.00

10413H03H5

DFS Item
 
life technologies
LS10413H03H5
5μg Each for $97.50

10413H03HL

DFS Item
 
life technologies
LS10413H03H250
5 x 50μg Each for $846.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias TNFRSF10D, CD264, DCR2, TRAILR4, TRUNDD

Tumor necrosis factor receptor superfamily, member 10D, decoy with truncated death domain (TNFRSF10D), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human TNFRSF10D-Fc Chimera at 10μg/mL (100μL/well) can bind biotinylated TNFSF10 with a linear range of 0.625-20ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.