missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ TNFRSF10D (TRAILR4) Recombinant Human Protein, hIgG1-Fc.His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-His 211) of human TNFRSF10D (NP_003831.2) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Manufacturer:  Life Technologies LS10413H03H50

 View more versions of this product

Catalog No. 10413H03H50


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; TNFRSF10D (TRAILR4) Recombinant Human Protein, hIgG1-Fc.His Tag; 50ug; Source: Human; Purity: greater than or equal to 90% by SDS-PAGE

Tumor necrosis factor receptor superfamily, member 10D, decoy with truncated death domain (TNFRSF10D), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human TNFRSF10D-Fc Chimera at 10μg/mL (100μL/well) can bind biotinylated TNFSF10 with a linear range of 0.625-20ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
44.7kDa
50μg
TNF Receptors
TNFRSF10D, CD264, DCR2, TRAILR4, TRUNDD
TNFRSF10D
Cytokines & Receptors/CD Proteins
His Tag/IgG1 Fc Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Tumor necrosis factor receptor superfamily, member 10d, decoy with truncated death domain
Assay Standard
Mammalian