Please login to your online account to display your discounted pricing

Sino Biological™ TNFRSF10B Recombinant Human Protein, hIgG1-Fc.His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain human TNFRSF10B (NP_003833.3) (Met 1-Glu 182) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Manufacturer: life technologies  LS10465H03H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10465H03H50

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Molecular Weight 50kDa
Purity ≥90% by SDS-PAGE
Validated Application Assay Standard
Protein Subtype TNF Receptors
Quantity 50μg
Protein Tag His Tag/IgG1 Fc Tag
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Common Name Tumor necrosis factor receptor superfamily, member 10b
Gene Alias CD262, DR5, KILLER, KILLER/DR5, TRAILR2, TRICK2, TRICK2A, TRICK2B, TRICKB, ZTNFR9
Expression System Mammalian
Gene Symbol TNFRSF10B
Protein Family Cytokines & Receptors/CD Proteins
Endotoxin Concentration <1 EU/μg
Format Lyophilized

TNFRSF10B, recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human TNFSF10 at 10μg/mL (100μL/well) can bind human TNFRSF10B-Fc chimera with a linear range of 2.5-40ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.