Please login to your online account to display your discounted pricing

Sino Biological™ TNF (TNFA) Recombinant Rat Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form of rat TNF-α (P 16599) (Val 80-Leu 235) fused to an initial Met at the N-terminus

$121.00 - $476.00

Specifications

Purity ≥90% by SDS-PAGE
Format Lyophilized
Gene Alias Tnf, mgC124630, RATTNF, TNF-alpha, Tnfa
View More Specs
Includes:  Five vials of lyophilized protein

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

80045RNAE25

DFS Item
 
life technologies
LS80045RNAE25
5 x 5μg Each for $278.00

80045RNAE5

DFS Item
 
life technologies
LS80045RNAE5
5μg Each for $121.00

80045RNAE50

DFS Item
 
life technologies
LS80045RNAE50
50μg Each for $476.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Format Lyophilized
Gene Alias Tnf, mgC124630, RATTNF, TNF-alpha, Tnfa

Tumor Necrosis Factor (TNF), recombinant rat protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

  • Activity: Measured in a cytotoxicity assay using L930 mouse fibrosarcoma cells in the presence of the metabolic inhibitor actinomycin D; the ED50 is typically 10-20pg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.