missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TNF (TNFA) Recombinant Cynomolgus Protein, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the soluble form of cynomolgus TNF-α (Val 77-Leu 233) fused to an initial Met at the N-terminus

$93.75 - $384.00

Specifications

Purity ≥90% by SDS-PAGE
Format Lyophilized
Gene Alias TNF-alpha
View More Specs

Includes: One vial of lyophilized protein

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
90018CNAE50 View Documents Life Technologies
LS90018CNAE50
50μg Each for $384.00
 
90018CNAE5 View Documents Life Technologies
LS90018CNAE5
5μg Each for $93.75
 
Description

Description

Tumor Necrosis Factor (TNF), recombinant cynomolgus protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured in a cytotoxicity assay using L-929 mouse fibrosarcoma cells in the presence of the metabolic inhibitor actinomycin D; the ED50 is typically 20-80pg/mL
  • Formulation: Lyophilized in 50mM Tris, 50mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
TNF-alpha

For Research Use Only. Not for use in diagnostic procedures.