Please login to your online account to display your discounted pricing

Sino Biological™ TNF (TNFA) Recombinant Canine Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding the soluble form of canine TNFα (NP_001003244.4) (Val 77-Leu 233) fused to an initial Met at the N-terminus

$135.00 - $326.00

Specifications

Purity ≥90% by SDS-PAGE
Format Frozen
Gene Alias TNFA, Ctnf
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

70003DNAE25

DFS Item
 
life technologies
LS70003DNAE25
5 x 5μg Each for $326.00

70003DNAE5

DFS Item
 
life technologies
LS70003DNAE5
5μg Each for $135.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Format Frozen
Gene Alias TNFA, Ctnf

Tumor Necrosis Factor (TNF), recombinant canine protein is supplied in frozen format. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

  • Activity: Measured in a cytotoxicity assay using L-929 mouse fibrosarcoma cells in the presence of the metabolic inhibitor actinomycin D; the ED50 is typically 0.2-1.2ng/mL
  • Formulation: 50mM Tris, 500mM NaCl, 20% glycerol, pH 8.0
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.