Please login to your online account to display your discounted pricing

Sino Biological™ TIMP2 Recombinant Human Protein DFS Item


This recombinant protein was expressed from a DNA sequence encoding the mature form of human TIMP2 (NP_003246.1) (Cys 27-Pro 220)

Manufacturer: life technologies  LS10396HNAH25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10396HNAH25

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 22kDa
Purity ≥95% by SDS-PAGE
Quantity 5 x 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Subtype TIMP
Endotoxin Concentration <1 EU/μg
Gene Alias CSC-21K, TIMP2
Common Name TIMP metallopeptidase inhibitor 2
Gene Symbol TIMP2
Protein Tag Untagged
Validated Application Assay Standard
Protein Family Protease Inhibitors
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized

TIMP Metallopeptidase Inhibitor 2 (TIMP2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to inhibit human mMP-2 cleavage of a fluorogenic peptide substrate MCA-PLGL-DPA-AR-NH2 (R&D Systems Cat. No. ES001), the IC50 value is approximately 2.2nM
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.