missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ TIMP1 Recombinant Human Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human TIMP1 (NP_003245.1) (Met 1-Ala 207)

Manufacturer:  Life Technologies LS10934H08H50

 View more versions of this product

Catalog No. 10934H08H50


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; TIMP1 Recombinant Human Protein; 50ug; Source: Human; Purity: greater than or equal to 90% by SDS-PAGE

TIMP metallopeptidase inhibitor 1 (TIMP1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to inhibit human mMP2 cleavage of a fluorogenic peptide substrate MCA-PLGL-DPA-AR-NH2, the IC50 value is approximately 6nM
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
22kDa
50μg
TIMP
TIMP1M, RP1-230G1.3, CLGI, EPA, EPO, FLJ90373, HCI, TIMP
TIMP1
Protease Inhibitors
Untagged
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
TIMP meallopeptidase inhibitor 1
Assay Standard
Mammalian