Please login to your online account to display your discounted pricing

Sino Biological™ TGM3 Recombinant Human Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human TGM3 (Q08188) (Ala 2-Glu 693) fused to a polyhistidine tag at the N-terminus

$199.50 - $199.50

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Frozen
Gene Alias MGC126249, mgC126250, TGE
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

11923H07B25

DFS Item
 
life technologies
LS11923H07B25
5 x 5μg Each for $199.50
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Frozen
Gene Alias MGC126249, mgC126250, TGE

Transglutaminase 3 (TGM3), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to cleave a synthetic peptide Benzyloxycarbonyl-Gln-Gly and NH2OH, the specific activity is >450pmole/min./μg
  • Formulation: 20mM Tris, 500mM NaCl, 0.2μg/μL BSA, pH 8.5, 10% glycerol
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.