Please login to your online account to display your discounted pricing

Sino Biological™ TGM3 Recombinant Human Protein DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human TGM3 (Q08188) (Ala 2-Glu 693) fused to a polyhistidine tag at the N-terminus

Manufacturer: life technologies  LS11923H07B25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11923H07B25

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 78.8kDa
Purity ≥90% by SDS-PAGE
Quantity 5 x 5μg
Storage Requirements Store at -80 °C. Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Termination N-Terminal
Protein Subtype Other Proteins
Endotoxin Concentration <1 EU/μg
Gene Alias MGC126249, mgC126250, TGE
Common Name Transglutaminase 3
Gene Symbol TGM3
Protein Tag His Tag
Validated Application Assay Standard
Expression System Insect
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Frozen

Transglutaminase 3 (TGM3), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to cleave a synthetic peptide Benzyloxycarbonyl-Gln-Gly and NH2OH, the specific activity is >450pmole/min./μg
  • Formulation: 20mM Tris, 500mM NaCl, 0.2μg/μL BSA, pH 8.5, 10% glycerol
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.